2,6-Difluoro-3-(methoxymethoxy)pyridine ≥95%

Supplier: AOBChem USA

Warning

30976-1G 30976-500MG 30976-5G
77468-818EA 168.92 USD
77468-818 77481-500 77491-884
2,6-Difluoro-3-(methoxymethoxy)pyridine ≥95%
2,6-Difluoro-3-(methoxymethoxy)pyridine

Precaución: For research use only

Formula: C₇H₇F₂NO₂
MW: 175.133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755892
CAS Number: 1638121-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR