α-Ethyl-2-fluoro-3-(trifluoromethoxy)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

309682-1G 309682-500MG 309682-5G
77468-814EA 415.09 USD
77468-814 77481-496 77491-880
α-Ethyl-2-fluoro-3-(trifluoromethoxy)benzenemethanol ≥95%
α-Ethyl-2-fluoro-3-(trifluoromethoxy)benzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₀F₄O₂
MW: 238.18 g/mol
Storage Temperature: Ambient
CAS Number: 1873936-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR