5-Bromo-2,3-difluoro-4-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

30946-1G 30946-500MG 30946-5G
77468-810EA 188.49 USD
77468-810 77481-492 77491-876
5-Bromo-2,3-difluoro-4-methylphenylboronic acid ≥97%
5-Bromo-2,3-difluoro-4-methylphenylboronic acid

Precaución: For research use only

Formula: C₇H₆BBrF₂O₂
MW: 250.833 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088231
CAS Number: 2121513-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR