2-(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

309214-1G 309214-500MG 309214-5G
77468-798EA 415.09 USD
77468-798 77481-480 77491-864
2-(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₉ClF₄O
Storage Temperature: Ambient
CAS Number: 3002428-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR