(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

309211-1G 309211-500MG 309211-5G
77468-794EA 415.09 USD
77468-794 77481-476 77491-860
(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)(isopropyl)sulfane ≥95%
(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₀H₉ClF₄S
MW: 272.6900 g/mol
Storage Temperature: Ambient
CAS Number: 3002530-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR