(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

309208-1G 309208-500MG 309208-5G
77468-790EA 415.09 USD
77468-790 77481-472 77491-856
(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)(ethyl)sulfane ≥95%
(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₇ClF₄S
Storage Temperature: Ambient
CAS Number: 3002519-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR