2-Chloro-1-(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)-2,2-difluoroethanone ≥95%

Warning

309206-1G 309206-500MG 309206-5G
77468-786 77481-468 77491-852
2-Chloro-1-(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₂Cl₂F₆O
MW: 311.0079 g/mol
Storage Temperature: Ambient
CAS Number: 3002467-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR