(3-(Benzyloxy)-6-bromo-2-fluorophenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

309201-1G 309201-500MG 309201-5G
77468-782EA 415.09 USD
77468-782 77481-464 77491-848
(3-(Benzyloxy)-6-bromo-2-fluorophenyl)(cyclopropyl)methanol ≥95%
(3-(Benzyloxy)-6-bromo-2-fluorophenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₇H₁₆BrFO₂
Storage Temperature: Ambient
CAS Number: 3002530-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR