1-(3-(Benzyloxy)-6-bromo-2-fluorophenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

309198-1G 309198-500MG 309198-5G
77468-778EA 415.09 USD
77468-778 77481-460 77491-844
1-(3-(Benzyloxy)-6-bromo-2-fluorophenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(3-(Benzyloxy)-6-bromo-2-fluorophenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₅H₉BrClF₃O₂
MW: 393.5829 g/mol
Storage Temperature: Ambient
CAS Number: 3002519-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR