2-(3-Methanesulfonyl-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%

Supplier: AOBChem USA

Warning

30916-1G 30916-500MG 30916-5G
77468-774EA 530.45 USD
77468-774 77481-456 77491-840
2-(3-Methanesulfonyl-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(3-Methanesulfonyl-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: For research use only

Formula: C₁₄H₂₁BO₅S
MW: 312.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22494013
CAS Number: 918328-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR