1-[3-[(4-Methylphenyl)methoxy]phenyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

308000-1G 308000-500MG 308000-5G
77468-766EA 515 USD
77468-766 77481-448 77491-832
1-[3-[(4-Methylphenyl)methoxy]phenyl]ethanone ≥95%
1-[3-[(4-Methylphenyl)methoxy]phenyl]ethanone

Precaución: For research use only

Formula: C₁₆H₁₆O₂
MW: 240.3 g/mol
Storage Temperature: Ambient
CAS Number: 202144-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR