2-(Cyclopropylmethoxy)-3,5-dimethylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Cyclopropylmethoxy)-3,5-dimethylbenzaldehyde

307966-1G 307966-500MG 307966-5G
77468-762EA 591.22 USD
77468-762 77481-444 77491-828
2-(Cyclopropylmethoxy)-3,5-dimethylbenzaldehyde ≥95%
2-(Cyclopropylmethoxy)-3,5-dimethylbenzaldehyde

Precaución: For research use only

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06246737
CAS Number: 883524-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR