1-Bromo-3-ethoxy-5-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

30773-1G 30773-500MG 30773-5G
77468-758EA 303.85 USD
77468-758 77481-440 77491-824
1-Bromo-3-ethoxy-5-methoxybenzene ≥95%
1-Bromo-3-ethoxy-5-methoxybenzene

Precaución: For research use only

Formula: C₉H₁₁BrO₂
MW: 231.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088203
CAS Number: 1235566-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR