5-Bromo-2,6-dimethoxypyridine-3-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30735-1G 30735-500MG 30735-5G
77468-754EA 130.81 USD
77468-754 77481-436 77491-820
5-Bromo-2,6-dimethoxypyridine-3-boronic acid pinacol ester ≥97%
5-Bromo-2,6-dimethoxypyridine-3-boronic acid pinacol ester

Precaución: For research use only

Formula: C₁₃H₁₉BBrNO₄
MW: 344.009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088184
CAS Number: 2121511-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR