Triethyl(2-fluoro-5-propoxyphenyl)silane ≥95%

Supplier: AOBChem USA

Warning

306994-1G 306994-500MG 306994-5G
77468-750EA 415.09 USD
77468-750 77481-432 77491-816
Triethyl(2-fluoro-5-propoxyphenyl)silane ≥95%
Triethyl(2-fluoro-5-propoxyphenyl)silane

Precaución: For research use only

Formula: C₁₅H₂₅FOSi
Storage Temperature: Ambient
MDL Number: MFCD34892840
CAS Number: 2969206-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR