1-(2-Fluoro-5-propoxyphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

306989-1G 306989-500MG 306989-5G
77468-746EA 415.09 USD
77468-746 77481-428 77491-812
1-(2-Fluoro-5-propoxyphenyl)cyclobutanol ≥95%
1-(2-Fluoro-5-propoxyphenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₃H₁₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD34892836
CAS Number: 2969382-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR