Ethyl(2-fluoro-5-propoxyphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

306987-1G 306987-500MG 306987-5G
77468-742EA 415.09 USD
77468-742 77481-424 77491-808
Ethyl(2-fluoro-5-propoxyphenyl)sulfane ≥95%
Ethyl(2-fluoro-5-propoxyphenyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₅FOS
MW: 214.2996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892834
CAS Number: 2969452-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR