2,2,3,3,3-pentafluoro-1-(2-fluoro-5-propoxyphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

306985-1G 306985-500MG 306985-5G
77468-738EA 415.09 USD
77468-738 77481-420 77491-804
2,2,3,3,3-pentafluoro-1-(2-fluoro-5-propoxyphenyl)propan-1-one ≥95%
2,2,3,3,3-pentafluoro-1-(2-fluoro-5-propoxyphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₂H₁₀F₆O₂
MW: 300.1970 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892832
CAS Number: 2969428-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR