2,2,2-trifluoro-1-(2-fluoro-5-propoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

306982-1G 306982-500MG 306982-5G
77468-734EA 415.09 USD
77468-734 77481-416 77491-800
2,2,2-trifluoro-1-(2-fluoro-5-propoxyphenyl)ethanone ≥95%
2,2,2-trifluoro-1-(2-fluoro-5-propoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₀F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34892829
CAS Number: 2969310-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR