(2-Fluoro-5-propoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

306980-1G 306980-500MG 306980-5G
77468-730EA 415.09 USD
77468-730 77481-412 77491-796
(2-Fluoro-5-propoxyphenyl)(methyl)sulfane ≥95%
(2-Fluoro-5-propoxyphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₃FOS
Storage Temperature: Ambient
MDL Number: MFCD34892827
CAS Number: 2969329-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR