4-Cyano-3-ethylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30629-1G 30629-500MG 30629-5G
77468-726EA 772.5 USD
77468-726 77481-408 77491-790
4-Cyano-3-ethylphenylboronic acid pinacol ester ≥97%
4-Cyano-3-ethylphenylboronic acid pinacol ester

Precaución: For research use only

Formula: C₁₅H₂₀BNO₂
MW: 257.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18732270
CAS Number: 2121513-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR