2-(4-Bromobenzo[b]thiophen-2-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

306254-1G 306254-500MG 306254-5G
77468-722EA 573.71 USD
77468-722 77481-404 77491-786
2-(4-Bromobenzo[b]thiophen-2-yl)propan-2-ol ≥95%
2-(4-Bromobenzo[b]thiophen-2-yl)propan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₁BrOS
MW: 271.1734 g/mol
Storage Temperature: Ambient
CAS Number: 1780932-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR