1-(4-Bromobenzo[b]thiophen-2-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

306250-1G 306250-500MG 306250-5G
77468-718EA 415.09 USD
77468-718 77481-400 77491-782
1-(4-Bromobenzo[b]thiophen-2-yl)cyclobutanol ≥95%
1-(4-Bromobenzo[b]thiophen-2-yl)cyclobutanol

Precaución: For research use only

Formula: C₁₂H₁₁BrOS
Storage Temperature: Ambient
CAS Number: 2250110-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR