(4-Bromobenzo[b]thiophen-2-yl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

306248-1G 306248-500MG 306248-5G
77468-714EA 415.09 USD
77468-714 77481-396 77491-778
(4-Bromobenzo[b]thiophen-2-yl)(cyclopropyl)methanol ≥95%
(4-Bromobenzo[b]thiophen-2-yl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₂H₁₁BrOS
MW: 283.1841 g/mol
Storage Temperature: Ambient
CAS Number: 3002435-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR