(4-Bromobenzo[b]thiophen-2-yl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

306246-1G 306246-500MG 306246-5G
77468-710EA 415.09 USD
77468-710 77481-392 77491-774
(4-Bromobenzo[b]thiophen-2-yl)triethylsilane ≥95%
(4-Bromobenzo[b]thiophen-2-yl)triethylsilane

Precaución: For research use only

Formula: C₁₄H₁₉BrSSi
MW: 327.3551 g/mol
Storage Temperature: Ambient
CAS Number: 3002473-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR