1-(4-Bromobenzo[b]thiophen-2-yl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

306244-1G 306244-500MG 306244-5G
77468-706EA 415.09 USD
77468-706 77481-388 77491-770
1-(4-Bromobenzo[b]thiophen-2-yl)-2-chloro-2,2-difluoroethanone ≥95%
1-(4-Bromobenzo[b]thiophen-2-yl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₄BrClF₂OS
MW: 325.5569 g/mol
Storage Temperature: Ambient
CAS Number: 3002426-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR