1-(4-Bromobenzo[b]thiophen-2-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

306242-1G 306242-500MG 306242-5G
77468-702EA 415.09 USD
77468-702 77481-384 77491-766
1-(4-Bromobenzo[b]thiophen-2-yl)-2,2-difluoroethanone ≥95%
1-(4-Bromobenzo[b]thiophen-2-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₅BrF₂OS
MW: 291.1119 g/mol
Storage Temperature: Ambient
CAS Number: 3002533-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR