5-Bromo-2-(ethylthio)benzofuran ≥95%

Supplier: AOBChem USA

Warning

306171-1G 306171-500MG 306171-5G
77468-698EA 415.09 USD
77468-698 77481-380 77491-762
5-Bromo-2-(ethylthio)benzofuran ≥95%
5-Bromo-2-(ethylthio)benzofuran

Precaución: For research use only

Formula: C₁₀H₉BrOS
MW: 257.1468 g/mol
Storage Temperature: Ambient
CAS Number: 3002538-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR