1-(5-Bromobenzofuran-2-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

306168-1G 306168-500MG 306168-5G
77468-694EA 415.09 USD
77468-694 77481-376 77491-758
1-(5-Bromobenzofuran-2-yl)-2,2-difluoroethanone ≥95%
1-(5-Bromobenzofuran-2-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₅BrF₂O₂
MW: 275.0463 g/mol
Storage Temperature: Ambient
CAS Number: 3002523-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR