2,5-Dibromobenzofuran ≥95%

Supplier: AOBChem USA

Warning

306166-1G 306166-500MG 306166-5G
77468-690EA 573.71 USD
77468-690 77481-372 77491-756
2,5-Dibromobenzofuran ≥95%
2,5-Dibromobenzofuran

Precaución: For research use only

Formula: C₈H₄Br₂O
Storage Temperature: Ambient
CAS Number: 1071108-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR