4-(4-Methylphenylsulfonamido)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

306082-1G 306082-5G
77468-682EA 60.77 USD
77468-682 77491-748
4-(4-Methylphenylsulfonamido)benzoic acid ≥95%
4-(4-Methylphenylsulfonamido)benzoic acid

Precaución: For research use only

Formula: C₁₄H₁₃NO₄S
MW: 291.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020311
CAS Number: 37028-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR