2,2-Difluoro-1-(5-methylbenzo[b]thiophen-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

306074-1G 306074-500MG 306074-5G
77468-678EA 415.09 USD
77468-678 77481-362 77491-744
2,2-Difluoro-1-(5-methylbenzo[b]thiophen-2-yl)ethanone ≥95%
2,2-Difluoro-1-(5-methylbenzo[b]thiophen-2-yl)ethanone

Precaución: For research use only

Formula: C₁₁H₈F₂OS
MW: 226.2424 g/mol
Storage Temperature: Ambient
CAS Number: 3002533-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR