Cyclopropyl(5-methylbenzo[b]thiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

306072-1G 306072-500MG 306072-5G
77468-674EA 415.09 USD
77468-674 77481-358 77491-740
Cyclopropyl(5-methylbenzo[b]thiophen-2-yl)methanol ≥95%
Cyclopropyl(5-methylbenzo[b]thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₃H₁₄OS
MW: 218.3146 g/mol
Storage Temperature: Ambient
CAS Number: 3002499-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR