1-(5-Bromobenzo[b]thiophen-2-yl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

306065-1G 306065-500MG 306065-5G
77468-670EA 415.09 USD
77468-670 77481-354 77491-736
1-(5-Bromobenzo[b]thiophen-2-yl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(5-Bromobenzo[b]thiophen-2-yl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₁H₄BrF₅OS
MW: 359.1098 g/mol
Storage Temperature: Ambient
CAS Number: 3002435-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR