5-Bromo-2-(ethylthio)benzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

306042-1G 306042-500MG 306042-5G
77468-666EA 415.09 USD
77468-666 77481-350 77491-732
5-Bromo-2-(ethylthio)benzo[b]thiophene ≥95%
5-Bromo-2-(ethylthio)benzo[b]thiophene

Precaución: For research use only

Formula: C₁₀H₉BrS₂
MW: 273.2124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892576
CAS Number: 2969315-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR