1-(5-Bromobenzo[b]thiophen-2-yl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

306040-1G 306040-500MG 306040-5G
77468-662EA 415.09 USD
77468-662 77481-346 77491-728
1-(5-Bromobenzo[b]thiophen-2-yl)-2-chloro-2,2-difluoroethanone ≥95%
1-(5-Bromobenzo[b]thiophen-2-yl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₄BrClF₂OS
MW: 325.5569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892574
CAS Number: 2969298-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR