2-(2-Methyl-5-propoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

306026-1G 306026-500MG 306026-5G
77468-654EA 415.09 USD
77468-654 77481-338 77491-720
2-(2-Methyl-5-propoxyphenyl)propan-2-ol ≥95%
2-(2-Methyl-5-propoxyphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
MW: 208.2967 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892570
CAS Number: 2969404-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR