1-(5-Methoxy-2-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

306015-1G 306015-500MG 306015-5G
77468-646EA 477.92 USD
77468-646 77481-330 77491-712
1-(5-Methoxy-2-methylphenyl)cyclobutanol ≥95%
1-(5-Methoxy-2-methylphenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
CAS Number: 2293746-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR