(5-Methoxy-2-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

306013-1G 306013-500MG 306013-5G
77468-642EA 415.09 USD
77468-642 77481-326 77491-708
(5-Methoxy-2-methylphenyl)(methyl)sulfane ≥95%
(5-Methoxy-2-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₂OS
MW: 168.2559 g/mol
Storage Temperature: Ambient
CAS Number: 82961-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR