Triethyl(5-methoxy-2-methylphenyl)silane ≥95%

Supplier: AOBChem USA

Warning

306010-1G 306010-500MG 306010-5G
77468-638EA 415.09 USD
77468-638 77481-322 77491-704
Triethyl(5-methoxy-2-methylphenyl)silane ≥95%
Triethyl(5-methoxy-2-methylphenyl)silane

Precaución: For research use only

Formula: C₁₄H₂₄OSi
Storage Temperature: Ambient
CAS Number: 3002492-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR