2-Chloro-2,2-difluoro-1-(5-methoxy-2-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

306008-1G 306008-500MG 306008-5G
77468-634EA 565.47 USD
77468-634 77481-318 77491-700
2-Chloro-2,2-difluoro-1-(5-methoxy-2-methylphenyl)ethanone ≥95%

Precaución: For research use only

Formula: C₁₀H₉ClF₂O₂
Storage Temperature: Ambient
CAS Number: 2293688-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR