4-Ethyl-4'-(methylthio)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

305932-1G 305932-500MG 305932-5G
77468-630EA 415.09 USD
77468-630 77481-314 77491-696
4-Ethyl-4'-(methylthio)-1,1'-biphenyl ≥95%
4-Ethyl-4'-(methylthio)-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₅H₁₆S
MW: 228.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892544
CAS Number: 104560-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR