[2-(Benzyloxy)pyridin-3-yl]methanol ≥97%

Supplier: AOBChem USA

Warning

30535-1G 30535-500MG 30535-5G
77468-626EA 197.76 USD
77468-626 77481-310 77491-692
[2-(Benzyloxy)pyridin-3-yl]methanol ≥97%
[2-(Benzyloxy)pyridin-3-yl]methanol

Precaución: For research use only

Formula: C₁₃H₁₃NO₂
MW: 215.248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14606377
CAS Number: 329980-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR