Methyl α-oxo-1-naphthaleneacetate ≥95%

Supplier: AOBChem USA

Warning

305271-1G 305271-500MG 305271-5G
77468-622EA 235.87 USD
77468-622 77481-306 77491-688
Methyl α-oxo-1-naphthaleneacetate ≥95%
Methyl α-oxo-1-naphthaleneacetate

Precaución: For research use only

Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26057481
CAS Number: 16738-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR