2-Ethoxy-3-pyridinemethanol ≥97%

Supplier: AOBChem USA

Warning

30526-1G 30526-500MG 30526-5G
77468-618EA 192.61 USD
77468-618 77481-302 77491-684
2-Ethoxy-3-pyridinemethanol ≥97%
2-Ethoxy-3-pyridinemethanol

Precaución: For research use only

Formula: C₈H₁₁NO₂
MW: 153.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14606402
CAS Number: 149489-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR