4-Chloro-2-fluoro-6-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

30459-1G 30459-500MG 30459-5G
77468-614EA 250.29 USD
77468-614 77481-298 77491-680
4-Chloro-2-fluoro-6-iodoanisole ≥97%
4-Chloro-2-fluoro-6-iodoanisole

Precaución: For research use only

Formula: C₇H₅ClFIO
MW: 286.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34181022
CAS Number: 1228093-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR