1-(4-(Benzyloxy)-3,5-dimethylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

303847-1G 303847-500MG 303847-5G
77468-610EA 415.09 USD
77468-610 77481-294 77491-676
1-(4-(Benzyloxy)-3,5-dimethylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(4-(Benzyloxy)-3,5-dimethylphenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₈H₁₅F₅O₂
MW: 358.3025 g/mol
Storage Temperature: Ambient
CAS Number: 3002525-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR