1-(4-(Benzyloxy)-3,5-dimethylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

303845-1G 303845-500MG 303845-5G
77468-606EA 415.09 USD
77468-606 77481-290 77491-672
1-(4-(Benzyloxy)-3,5-dimethylphenyl)-2,2-difluoroethanone ≥95%
1-(4-(Benzyloxy)-3,5-dimethylphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₇H₁₆F₂O₂
MW: 290.3045 g/mol
Storage Temperature: Ambient
CAS Number: 3002437-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR