(4-Isopropoxy-2-methylphenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

303842-1G 303842-500MG 303842-5G
77468-602EA 415.09 USD
77468-602 77481-286 77491-668
(4-Isopropoxy-2-methylphenyl)(isopropyl)sulfane ≥95%
(4-Isopropoxy-2-methylphenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₃H₂₀OS
Storage Temperature: Ambient
CAS Number: 1081546-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR