Ethyl(4-isopropoxy-2-methylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

303833-1G 303833-500MG 303833-5G
77468-598EA 415.09 USD
77468-598 77481-282 77491-664
Ethyl(4-isopropoxy-2-methylphenyl)sulfane ≥95%
Ethyl(4-isopropoxy-2-methylphenyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₈OS
Storage Temperature: Ambient
MDL Number: MFCD34892147
CAS Number: 2969403-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR